3-Methylthiophene 99%
Supplier: Apollo Scientific
|
Danger
|
OR10137-25G
OR10137-100G
OR10137-500G
APOSOR10137-25GEA
17.6
EUR
APOSOR10137-25G
APOSOR10137-100G
APOSOR10137-500G
3-Methylthiophene 99%
3-Methylthiophene
Formula:
C₅H₆S MW: 98,17 g/mol Boiling Pt: 115…117 °C Melting Pt: –69 °C Density: 1,016 g/cm³ (25 °C) Flash Pt: 11 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005470 CAS Number: 616-44-4 EINECS: 210-482-6 UN: 1993 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...