2-Acetylbenzo[b]thiophene 98%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
2-Acetylbenzothiophene
OR10146-100G
OR10146-25G
APOSOR10146-100GEA
125
EUR
APOSOR10146-100G
APOSOR10146-25G
2-Acetylbenzo[b]thiophene 98%
2-Acetylbenzo[b]thiophene
Formula:
C₁₀H₈OS MW: 176.23 g/mol Melting Pt: 89…90 °C Storage Temperature: Ambient |
MDL Number:
MFCD00090217 CAS Number: 22720-75-8 UN: 3077 ADR: 9+EHS III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...