6-Nitroindole
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
6-Nitro-1H-indole
OR10187-1G
OR10187-5G
APOSOR10187-1GEA
21.9
EUR
APOSOR10187-1G
APOSOR10187-5G
6-Nitroindole
6-Nitroindole
Formula:
C₈H₆N₂O₂ MW: 162,15 g/mol Melting Pt: 137…143 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00051497 CAS Number: 4769-96-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...