4,6-Dichloro-3-(4-chlorophenyl)pyridazine 95+%

Supplier: Apollo Scientific

OR1021-1G OR1021-5G OR1021-10G
APOSOR1021-1GEA 260 EUR
APOSOR1021-1G APOSOR1021-5G APOSOR1021-10G
4,6-Dichloro-3-(4-chlorophenyl)pyridazine 95+%
4,6-Dichloro-3-(4-chlorophenyl)pyridazine
Formula: C₁₀H₅Cl₃N₂
MW: 259.52 g/mol
Melting Pt: 163…165 °C
Storage Temperature: Ambient
MDL Number: MFCD06800550
CAS Number: 68240-46-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR