4-Aminotetrahydropyran
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
Tetrahydro-2H-pyran-4-amine
OR10342-5G
OR10342-1G
OR10342-25G
APOSOR10342-5GEA
24.8
EUR
APOSOR10342-5G
APOSOR10342-1G
APOSOR10342-25G
4-Aminotetrahydropyran
4-Aminotetrahydropyran
Formula:
C₅H₁₁NO MW: 101,1469 g/mol Boiling Pt: 45…55 °C/20mmHg Density: 0,977 g/cm³ (25 °C) Flash Pt: 54,4 °C Storage Temperature: Ambient |
MDL Number:
MFCD02179436 CAS Number: 38041-19-9 UN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...