5-Bromo-1-methylimidazole
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
5-Bromo-1-methyl-1H-imidazole
OR10368-25G
OR10368-5G
OR10368-100G
APOSOR10368-25GEA
63.8
EUR
APOSOR10368-25G
APOSOR10368-5G
APOSOR10368-100G
5-Bromo-1-methylimidazole
5-Bromo-1-methylimidazole
Formula:
C₄H₅BrN₂ MW: 161 g/mol Boiling Pt: 120 °C/15mmHg Melting Pt: 40…41 °C Flash Pt: 82 °C Storage Temperature: Ambient |
MDL Number:
MFCD01632218 CAS Number: 1003-21-0 UN: 3335 ADR: 9,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...