4-(tert-Butoxycarboxy)phenylboronic acid 98%

Supplier: Apollo Scientific
Warning

OR10395-1G OR10395-5G
APOSOR10395-1GEA 107 EUR
APOSOR10395-1G APOSOR10395-5G
4-(tert-Butoxycarboxy)phenylboronic acid 98%
4-(tert-Butoxycarboxy)phenylboronic acid
Formula: C₁₁H₁₅BO₅
MW: 238,05 g/mol
Melting Pt: 140…146 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03095144
CAS Number: 380430-70-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR