4,5-Diiodoimidazole 98%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
4,5-Diiodo-1H-imidazole
OR10450-1G
OR10450-5G
APOSOR10450-1GEA
21.9
EUR
APOSOR10450-1G
APOSOR10450-5G
4,5-Diiodoimidazole 98%
4,5-Diiodoimidazole
Formula:
C₃H₂I₂N₂ MW: 319,87 g/mol Melting Pt: 180…181 °C Storage Temperature: Ambient |
MDL Number:
MFCD00963878 CAS Number: 15813-09-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...