3-Hydroxyphenylboronic acid 98%

Supplier: Apollo Scientific
Warning

Synonyms: (3-Hydroxyphenyl)metaboric acid, (3-Hydroxyphenyl)boronic acid, Dihydroxy(3-hydroxyphenyl)borane, 3-Hydroxybenzeneboronic acid

OR10504-25G OR10504-5G
APOSOR10504-25GEA 79.9 EUR
APOSOR10504-25G APOSOR10504-5G
3-Hydroxyphenylboronic acid 98%
3-Hydroxyphenylboronic acid
Formula: C₆H₇BO₃
MW: 137,93 g/mol
Melting Pt: 208…213 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD01074603
CAS Number: 87199-18-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR