5-(Hydroxymethyl)indole
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
(1H-Indol-5-yl)methanol
OR10507-1G
OR10507-250MG
OR10507-5G
APOSOR10507-1GEA
119
EUR
APOSOR10507-1G
APOSOR10507-250MG
APOSOR10507-5G
5-(Hydroxymethyl)indole
5-(Hydroxymethyl)indole
Formula:
C₉H₉NO MW: 147,1700 g/mol Melting Pt: 66…70 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD02179594 CAS Number: 1075-25-8 |
Learn more
![](/m-ie.vwr.com/en_IE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...