3-Isopropylphenylboronic acid 98%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
3-Isopropylbenzeneboronic acid
OR10523-10G
OR10523-5G
APOSOR10523-10GEA
198
EUR
APOSOR10523-10G
APOSOR10523-5G
3-Isopropylphenylboronic acid 98%
3-Isopropylphenylboronic acid
Formula:
C₉H₁₃BO₂ MW: 164,01 g/mol Melting Pt: 72…76 °C Flash Pt: >110 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD01074634 CAS Number: 216019-28-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...