Methyl-2-aminothiazole-5-carboxylate

Supplier: Apollo Scientific
Warning

Synonyms: Methyl 2-aminothiazole-5-carboxylate, 2-Aminothiazole-5-carboxylic acid methyll ester

OR10538-25G OR10538-5G
APOSOR10538-25GEA 85.8 EUR
APOSOR10538-25G APOSOR10538-5G
Methyl-2-aminothiazole-5-carboxylate
Methyl-2-aminothiazole-5-carboxylate
Formula: C₅H₆N₂O₂S
MW: 158,1800 g/mol
Melting Pt: 191 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03788562
CAS Number: 6633-61-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR