4-(Methylsulphinylphenyl)boronic acid

Supplier: Apollo Scientific

Synonyms: (4-(Methylsulphinyl)phenyl)boronic acid

OR10559-1G OR10559-100MG OR10559-250MG OR10559-500MG
APOSOR10559-1GEA 353 EUR
APOSOR10559-1G APOSOR10559-100MG APOSOR10559-250MG APOSOR10559-500MG
4-(Methylsulphinylphenyl)boronic acid
4-(Methylsulphinylphenyl)boronic acid
Formula: C₇H₉BO₃S
MW: 184,02 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02093071
CAS Number: 166386-48-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR