5-Nitrooxindole 98%

Supplier: Apollo Scientific
Warning

Synonyms: 5-Nitroindolin-2-one, 5-Nitro-2-oxindole

OR10569-100G OR10569-10G OR10569-25G
APOSOR10569-100GEA 741 EUR
APOSOR10569-100G APOSOR10569-10G APOSOR10569-25G
5-Nitrooxindole 98%
5-Nitrooxindole
Formula: C₈H₆N₂O₃
MW: 178.15 g/mol
Melting Pt: 233…236 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00456999
CAS Number: 20870-79-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR