1-Tritylimidazole-4-carbaldehyde 95%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
1-Trityl-1H-imidazole-4-carboxaldehyde, 1-Trityl-1H-imidazole-4-carbaldehyde, 1-Tritylimidazole-4-carboxaldehyde
OR10596-5G
OR10596-10G
OR10596-25G
APOSOR10596-5GEA
21.9
EUR
APOSOR10596-5G
APOSOR10596-10G
APOSOR10596-25G
1-Tritylimidazole-4-carbaldehyde 95%
1-Tritylimidazole-4-carbaldehyde
Formula:
C₂₃H₁₈N₂O MW: 338,41 g/mol Melting Pt: 187…192 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD02179554 CAS Number: 33016-47-6 |
Learn more
![](/m-ie.vwr.com/en_IE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...