4-Vinylphenylboronic acid 97%

Supplier: Apollo Scientific
Warning

Synonyms: Dihydroxy(4-vinylphenyl)borane, (4-Vinylphenyl)metaboric acid, 4-Vinylbenzeneboronic acid

OR10599-25G OR10599-5G
APOSOR10599-25GEA 270 EUR
APOSOR10599-25G APOSOR10599-5G
4-Vinylphenylboronic acid 97%
4-Vinylphenylboronic acid
Formula: C₈H₉BO₂
MW: 147,97 g/mol
Melting Pt: 190…193 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00239441
CAS Number: 2156-04-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR