Bis(neopentyl glycolato)diboron
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
5,5,5',5'-Tetramethyl-2,2'-bi(1,3,2-dioxaborinane), 2-(5,5-dimethyl-1,3,2-dioxaborinan-2-yl)-5,5-dimethyl-1,3,2-dioxaborinane
OR10607-100G
OR10607-25G
OR10607-500G
APOSOR10607-100GEA
52.2
EUR
APOSOR10607-100G
APOSOR10607-25G
APOSOR10607-500G
Bis(neopentyl glycolato)diboron
Bis(neopentyl glycolato)diboron
Formula:
C₁₀H₂₀B₂O₄ MW: 225,89 g/mol Melting Pt: 175…185 °C Storage Temperature: Ambient |
MDL Number:
MFCD02093062 CAS Number: 201733-56-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...