4-Bromoaniline 98%
Supplier: Apollo Scientific
|
Danger
|
OR10617-100G
OR10617-25G
APOSOR10617-100GEA
78.3
EUR
APOSOR10617-100G
APOSOR10617-25G
4-Bromoaniline 98%
4-Bromoaniline
Formula:
C₆H₆BrN MW: 172,02 g/mol Boiling Pt: 222…224 °C Melting Pt: 56…65 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007822 CAS Number: 106-40-1 EINECS: 203-393-9 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...