4-Bromophenol 98%

Supplier: Apollo Scientific
Warning

OR10618-25G OR10618-100G OR10618-500G
APOSOR10618-25GEA 21.9 EUR
APOSOR10618-25G APOSOR10618-100G APOSOR10618-500G
4-Bromophenol 98%
4-Bromophenol
Formula: C₆H₅BrO
MW: 173.01 g/mol
Boiling Pt: 235…238 °C
Melting Pt: 61…66 °C
Density: 1.84 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00002313
CAS Number: 106-41-2
EINECS: 203-394-4
UN: 3077
ADR: 9+EHS III
Merck Index: 13,01413

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR