Benzyl dimethylphosphonoacetate 98%

Supplier: Apollo Scientific
Warning

OR10755-1G OR10755-5G
APOSOR10755-1GEA 98.9 EUR
APOSOR10755-1G APOSOR10755-5G
Benzyl dimethylphosphonoacetate 98%
Benzyl dimethylphosphonoacetate
Formula: C₁₁H₁₅O₅P
Boiling Pt: 148…150 °C/0,1mmHg
Density: 1,23 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD01872507
CAS Number: 57443-18-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR