6-Bromo-2-tetralone 97%

Supplier: Apollo Scientific
Warning

Synonyms: 6-Bromo-3,4-dihydronaphthalen-2(1H)-one

OR10839-1G OR10839-5G
APOSOR10839-1GEA 72.7 EUR
APOSOR10839-1G APOSOR10839-5G
6-Bromo-2-tetralone 97%
6-Bromo-2-tetralone
Formula: C₁₀H₉BrO
MW: 225,08 g/mol
Melting Pt: 73…77 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00239388
CAS Number: 4133-35-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR