2,5-Dichlorotoluene 98%

Supplier: Apollo Scientific
Warning

Synonyms: 1,4-Dichloro-2-methylbenzene

OR1084-25G OR1084-100G
APOSOR1084-25GEA 69.8 EUR
APOSOR1084-25G APOSOR1084-100G
2,5-Dichlorotoluene 98%
2,5-Dichlorotoluene
Formula: C₇H₆Cl₂
MW: 161,03 g/mol
Boiling Pt: 197…200 °C
Melting Pt: 44320 °C
Density: 1,256 g/cm³ (25 °C)
Flash Pt: 79 °C
Storage Temperature: Ambient
MDL Number: MFCD00000609
CAS Number: 19398-61-9
EINECS: 243-032-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR