2-Chloro-2-ethoxyacetic acid ethyl ester

Supplier: Apollo Scientific
Danger

OR10840-5G OR10840-1G
APOSOR10840-5GEA 144 EUR
APOSOR10840-5G APOSOR10840-1G
2-Chloro-2-ethoxyacetic acid ethyl ester
2-Chloro-2-ethoxyacetic acid ethyl ester
Formula: C₆H₁₁ClO₃
MW: 166,601 g/mol
Boiling Pt: 81…82 °C/12mmHg
Storage Temperature: Ambient
MDL Number: MFCD00043953
CAS Number: 34006-60-5
EINECS: 251-786-9
UN: 1760
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR