Ethyl-1-benzofuran-2-carboxylate 97%

Supplier: Apollo Scientific
Warning

Synonyms: Ethyl benzofuran-2-carboxylate

OR10935-1G OR10935-5G
APOSOR10935-1GEA 0 EUR
APOSOR10935-1G APOSOR10935-5G
Ethyl-1-benzofuran-2-carboxylate 97%
Ethyl-1-benzofuran-2-carboxylate
Formula: C₁₁H₁₀O₃
MW: 190,2 g/mol
Melting Pt: 29…34 °C
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03094950
CAS Number: 3199-61-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR