5-Chlorooxindole 98%

Supplier: Apollo Scientific
Warning

Synonyms: 5-Chloro-2-oxindole

OR10940-5G OR10940-100G OR10940-25G
APOSOR10940-5GEA 62.5 EUR
APOSOR10940-5G APOSOR10940-100G APOSOR10940-25G
5-Chlorooxindole 98%
5-Chlorooxindole
Formula: C₈H₆ClNO
MW: 167,59 g/mol
Melting Pt: 194…200 °C
Storage Temperature: Ambient
MDL Number: MFCD00191927
CAS Number: 17630-75-0
EINECS: 412-200-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR