5-Chloro-2-benzothiazolinone
Supplier: Apollo Scientific
|
Warning
|
OR10944-25G
OR10944-5G
APOSOR10944-25GEA
81.2
EUR
APOSOR10944-25G
APOSOR10944-5G
5-Chloro-2-benzothiazolinone
5-Chloro-2-benzothiazolinone
Formula:
C₇H₄ClNOS Melting Pt: 237 °C Storage Temperature: Ambient |
MDL Number:
MFCD00060125 CAS Number: 20600-44-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...