Methyl-4-bromocrotonate 80%

Supplier: Apollo Scientific
Danger

Synonyms: Methyl 4-bromobut-2-enoate

OR10966-100G OR10966-25G
APOSOR10966-100GEA 234 EUR
APOSOR10966-100G APOSOR10966-25G
Methyl-4-bromocrotonate 80%
Methyl 4-bromocrotonate
Formula: C₅H₇BrO₂
MW: 179,01 g/mol
Boiling Pt: 83…85 °C/13mmHg
Density: 1,522 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00000246
CAS Number: 1117-71-1
UN: 3334
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR