(4-Bromo-3,5-dimethyl-1H-pyrazol-1-yl)methanol

Supplier: Apollo Scientific
Warning

OR110037-10G OR110037-1G OR110037-5G
APOSOR110037-10GEA 350 EUR
APOSOR110037-10G APOSOR110037-1G APOSOR110037-5G
(4-Bromo-3,5-dimethyl-1H-pyrazol-1-yl)methanol
(4-Bromo-3,5-dimethyl-1H-pyrazol-1-yl)methanol
Formula: C₆H₉BrN₂O
Storage Temperature: Ambient
MDL Number: MFCD00085534
CAS Number: 94230-83-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR