Ethyl-3-amino-5-chlorobenzofuran-2-carboxylate

Supplier: Apollo Scientific
Warning

OR110056-1G OR110056-250MG
APOSOR110056-1GEA 209 EUR
APOSOR110056-1G APOSOR110056-250MG
Ethyl-3-amino-5-chlorobenzofuran-2-carboxylate
Ethyl-3-amino-5-chlorobenzofuran-2-carboxylate
Formula: C₁₁H₁₀ClNO₃
MW: 239,66 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00454352
CAS Number: 329210-07-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR