2-Butyl-6-nitrobenzoxazole
Supplier: Apollo Scientific
OR110139-10G
OR110139-1G
OR110139-5G
APOSOR110139-10GEA
372
EUR
APOSOR110139-10G
APOSOR110139-1G
APOSOR110139-5G
2-Butyl-6-nitrobenzoxazole
2-Butyl-6-nitrobenzoxazole
Formula:
C₁₁H₁₂N₂O₃ Storage Temperature: Ambient |
MDL Number:
MFCD05663809 CAS Number: 886360-96-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...