1-Methyl-1H-pyrazole
Supplier: Apollo Scientific
Synonyms:
1-Methylpyrazole
OR110194-10G
OR110194-1G
OR110194-5G
APOSOR110194-10GEA
0
EUR
APOSOR110194-10G
APOSOR110194-1G
APOSOR110194-5G
1-Methyl-1H-pyrazole
1-Methyl-1H-pyrazole
Formula:
C₄H₆N₂ MW: 82,11 g/mol Boiling Pt: 127 °C Density: 0,988 g/cm³ (25 °C) Flash Pt: 36 °C Storage Temperature: Ambient |
MDL Number:
MFCD00144943 CAS Number: 930-36-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...