Ethyl-5-nitrobenzo[b]thiophene-2-carboxylate

Supplier: Apollo Scientific

OR110195-1G OR110195-5G
APOSOR110195-1GEA 46.6 EUR
APOSOR110195-1G APOSOR110195-5G
Ethyl-5-nitrobenzo[b]thiophene-2-carboxylate
Ethyl 5-nitrobenzo[b]thiophene-2-carboxylate
Formula: C₁₁H₉NO₄S
MW: 251,258 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03791163
CAS Number: 25785-09-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR