5-Nitro-1-benzofuran-2-carbohydrazide

Supplier: Apollo Scientific
Warning

OR110200-1G OR110200-5G
APOSOR110200-1GEA 58 EUR
APOSOR110200-1G APOSOR110200-5G
5-Nitro-1-benzofuran-2-carbohydrazide
5-Nitro-1-benzofuran-2-carbohydrazide
Formula: C₉H₇N₃O₄
MW: 221,17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11553052
CAS Number: 406192-63-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR