2-Propyl-1H-benzo[d]imidazole

Supplier: Apollo Scientific
Warning

Synonyms: 2-Propylbenzimidazole

OR110235-5G OR110235-10G
APOSOR110235-5GEA 121 EUR
APOSOR110235-5G APOSOR110235-10G
2-Propyl-1H-benzo[d]imidazole
2-Propyl-1H-benzo[d]imidazole
Formula: C₁₀H₁₂N₂
MW: 160,2200 g/mol
Melting Pt: 150…153 °C
Storage Temperature: Ambient
MDL Number: MFCD00087477
CAS Number: 5465-29-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR