2-Methyl-1,3-benzoxazole-5-carboxylic acid 95%

Supplier: Apollo Scientific
Warning

Synonyms: 2-Methylbenzo[d]oxazole-5-carboxylic acid

OR110248-5G
APOSOR110248-5GEA 85.7 EUR
APOSOR110248-5G
2-Methyl-1,3-benzoxazole-5-carboxylic acid 95%
2-Methyl-1,3-benzoxazole-5-carboxylic acid
Formula: C₉H₇NO₃
MW: 177,16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09759125
CAS Number: 90322-32-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR