Methyl-2-amino-5-bromonicotinate

Supplier: Apollo Scientific
Warning

Synonyms: Methyl 2-amino-5-bromopyridine-3-carboxylate

OR110269-1G OR110269-5G
APOSOR110269-1GEA 51 EUR
APOSOR110269-1G APOSOR110269-5G
Methyl-2-amino-5-bromonicotinate
Methyl 2-amino-5-bromonicotinate
Formula: C₇H₇BrN₂O₂
MW: 231,05 g/mol
Melting Pt: 145…146 °C
Storage Temperature: Ambient
MDL Number: MFCD04117787
CAS Number: 50735-34-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR