2,6-Dichloropyrazine 98%
Supplier: Apollo Scientific
|
Warning
|
OR11034-100G
OR11034-250G
OR11034-25G
APOSOR11034-100GEA
26.1
EUR
APOSOR11034-100G
APOSOR11034-250G
APOSOR11034-25G
2,6-Dichloropyrazine 98%
2,6-Dichloropyrazine
Formula:
C₄H₂Cl₂N₂ MW: 148,98 g/mol Melting Pt: 53…56 °C Flash Pt: 113 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006125 CAS Number: 4774-14-5 EINECS: 225-316-8 |
Learn more
![](/m-ie.vwr.com/en_IE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...