Methyl-5-chloro-6-oxo-1,6-dihydro-3-pyridinecarboxylate

Supplier: Apollo Scientific
Warning

Synonyms: Methyl 5-chloro-6-hydroxynicotinate, Methyl 5-chloro-6-oxo-1,6-dihydropyridine-3-carboxylate

OR110393-1G OR110393-5G
APOSOR110393-1GEA 132 EUR
APOSOR110393-1G APOSOR110393-5G
Methyl-5-chloro-6-oxo-1,6-dihydro-3-pyridinecarboxylate
Methyl-5-chloro-6-oxo-1,6-dihydro-3-pyridinecarboxylate
Formula: C₇H₆ClNO₃
MW: 187,58 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD03844853
CAS Number: 316166-47-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR