Methyl [4-(3-oxobutyl)phenoxy]acetate

Supplier: Apollo Scientific

OR110461-1G OR110461-5G
APOSOR110461-1GEA 87 EUR
APOSOR110461-1G APOSOR110461-5G
Methyl [4-(3-oxobutyl)phenoxy]acetate
Methyl [4-(3-oxobutyl)phenoxy]acetate
Formula: C₁₃H₁₆O₄
Storage Temperature: Ambient
MDL Number: MFCD09933374
CAS Number: 78069-49-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR