2-Chloro-5-methyl-3-nitropyridine 97%

Supplier: Apollo Scientific
Warning

Synonyms: 2-Chloro-3-nitro-5-picoline

OR11054-100G OR11054-25G OR11054-5G
APOSOR11054-100GEA 114 EUR
APOSOR11054-100G APOSOR11054-25G APOSOR11054-5G
2-Chloro-5-methyl-3-nitropyridine 97%
2-Chloro-5-methyl-3-nitropyridine
Formula: C₆H₅ClN₂O₂
MW: 172.57 g/mol
Melting Pt: 46…48 °C
Storage Temperature: Ambient
MDL Number: MFCD02070020
CAS Number: 23056-40-8
UN: 2811
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR