6-Bromo-2H-chromene-3-carboxylic acid
Supplier: Apollo Scientific
OR110565-1G
OR110565-5G
OR110565-250MG
APOSOR110565-1GEA
132
EUR
APOSOR110565-1G
APOSOR110565-5G
APOSOR110565-250MG
6-Bromo-2H-chromene-3-carboxylic acid
6-Bromo-2H-chromene-3-carboxylic acid
Formula:
C₁₀H₇BrO₃ Storage Temperature: Ambient |
MDL Number:
MFCD04035698 CAS Number: 380607-15-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...