4'-Hydroxypropiophenone 98%

Supplier: Apollo Scientific

Synonyms: 4’-Hydroxypropiophenone, Paroxypropione, p-Hydroxypropiophenone

OR11062-10KG OR11062-1KG OR11062-250G
APOSOR11062-10KGEA 1430 EUR
APOSOR11062-10KG APOSOR11062-1KG APOSOR11062-250G
4'-Hydroxypropiophenone 98%
4'-Hydroxypropiophenone
Formula: C₉H₁₀O₂
MW: 150,18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00002361
CAS Number: 70-70-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR