4'-Methoxyacetophenone 99%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
Acetanisole, 4-Methoxyacetophenone
OR11063-1KG
OR11063-250G
APOSOR11063-1KGEA
194
EUR
APOSOR11063-1KG
APOSOR11063-250G
4'-Methoxyacetophenone 99%
4'-Methoxyacetophenone
Formula:
C₉H₁₀O₂ MW: 150,18 g/mol Boiling Pt: 152…154 °C/26mmHg Melting Pt: 36…38 °C Density: 1,08 g/cm³ (25 °C) Flash Pt: 138 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008745 CAS Number: 100-06-1 EINECS: 202-815-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...