4-Amino-1-methyl-1H-pyrazole 95%

Supplier: Apollo Scientific
Warning

Synonyms: 1-methyl-1H-pyrazol-4-amine

OR110642-100G OR110642-25G
APOSOR110642-100GEA 244 EUR
APOSOR110642-100G APOSOR110642-25G
4-Amino-1-methyl-1H-pyrazole 95%
4-Amino-1-methyl-1H-pyrazole
Formula: C₄H₇N₃
MW: 97,12 g/mol
Boiling Pt: 110…112 °C/2mmHg
Storage Temperature: Ambient
MDL Number: MFCD05667135
CAS Number: 69843-13-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR