ethyl-2-((tert-butoxycarbonyl)amino)acetate
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
N-(tert-Butoxycarbonyl)glycine ethyl ester, ethyl 2-((tert-Butoxycarbonyl)amino)acetate, Boc-Gly-OEt, N-Boc-glycine ethyl ester
OR110664-100G
OR110664-25G
OR110664-5G
APOSOR110664-100GEA
389
EUR
APOSOR110664-100G
APOSOR110664-25G
APOSOR110664-5G
ethyl-2-((tert-butoxycarbonyl)amino)acetate
ethyl-2-((tert-butoxycarbonyl)amino)acetate
Formula:
C₉H₁₇NO₄ MW: 203.2 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD05663720 CAS Number: 14719-37-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...