Ethyl-3-chloroquinoxaline-2-carboxylate
Supplier: Apollo Scientific
|
Warning
|
OR110667-1G
OR110667-5G
APOSOR110667-1GEA
65.4
EUR
APOSOR110667-1G
APOSOR110667-5G
Ethyl-3-chloroquinoxaline-2-carboxylate
Ethyl-3-chloroquinoxaline-2-carboxylate
Formula:
C₁₁H₉ClN₂O₂ Melting Pt: 33…36 °C Storage Temperature: Ambient |
MDL Number:
MFCD09953512 CAS Number: 49679-45-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...