7-Chloro-5-nitro-1H-indazole
Supplier: Apollo Scientific
|
Warning
|
OR110736-1G
OR110736-250MG
APOSOR110736-1GEA
261
EUR
APOSOR110736-1G
APOSOR110736-250MG
7-Chloro-5-nitro-1H-indazole
7-Chloro-5-nitro-1H-indazole
Formula:
C₇H₄ClN₃O₂ Melting Pt: 240…243 °C Storage Temperature: Ambient |
CAS Number:
74209-22-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...