2-(Chloromethyl)-5-nitrobenzoxazole

Supplier: Apollo Scientific
Warning

OR110829-1G OR110829-5G
APOSOR110829-1GEA 288 EUR
APOSOR110829-1G APOSOR110829-5G
2-(Chloromethyl)-5-nitrobenzoxazole
2-(Chloromethyl)-5-nitrobenzoxazole
Formula: C₈H₅ClN₂O₃
Storage Temperature: Ambient
MDL Number: MFCD01596292
CAS Number: 119198-10-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR