2-(1-Oxo-1,3-dihydro-2H-isoindol-2-yl)propanoic acid

Supplier: Apollo Scientific

OR110883-1G OR110883-5G
APOSOR110883-1GEA 58.2 EUR
APOSOR110883-1G APOSOR110883-5G
2-(1-Oxo-1,3-dihydro-2H-isoindol-2-yl)propanoic acid
2-(1-Oxo-1,3-dihydro-2H-isoindol-2-yl)propanoic acid
Formula: C₁₁H₁₁NO₃
Melting Pt: 202…204 °C
Storage Temperature: Ambient
MDL Number: MFCD00664643
CAS Number: 67266-14-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR