3-(1-Oxo-1,3-dihydro-2H-isoindol-2-yl)propanoic acid

Supplier: Apollo Scientific
Warning

OR110885-1G OR110885-5G
APOSOR110885-1GEA 116 EUR
APOSOR110885-1G APOSOR110885-5G
3-(1-Oxo-1,3-dihydro-2H-isoindol-2-yl)propanoic acid
3-(1-Oxo-1,3-dihydro-2H-isoindol-2-yl)propanoic acid
Formula: C₁₁H₁₁NO₃
Melting Pt: 113…116 °C
Storage Temperature: Ambient
MDL Number: MFCD03131298
CAS Number: 83747-30-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR